missing translation for 'onlineSavingsMsg'
Learn More

GLIPR1L1 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18410338 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410338 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410338 Supplier Novus Biologicals Supplier No. NBP281989V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLIPR1L1 Polyclonal antibody specifically detects GLIPR1L1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIPR1L1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias ALKN2972, GLI pathogenesis-related 1 like 1, GLIPR1-like protein 1, MGC26856, PRO7434
Host Species Rabbit
Immunogen GLIPR1L1 antibody was raised against an 18 amino acid peptide near the amino terminus of human GLIPR1L1 . The immunogen is located within amino acids 20 - 70 of GLIPR1L1. Amino Acid Squence: IEAHNEWRGKVNPPAAD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256710
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.