missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLIPR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595776
This item is not returnable.
View return policy
Description
Immunogen sequence: ANILPDIENE DFIKDCVRIH NKFRSEVKPT ASDMLYMTWD PALAQIAKAW ASNCQFSHNT RLKPPHKLHP NFTSLGENIW TGSVPIFSVS SAITNWYDEI QDYDFKTRIC KKVCGHYTQ; Positive Samples: Rat lung; Cellular Location: Membrane, Single-pass membrane protein.
The glioma pathogensis-related protein 1 (GLIPR1) is similar to both the pathogenesis-related protein (PR) superfamily and the cysteine-rich secretory protein (CRISP) family. GLIPR1 is a tumor suppressor whose expression is regulated by p53; its increased expression is associated with myelomocytic differentiation in macrophages, whereas decreased expression of this gene through gene methylation is associated with prostate cancer. The GLIPR1 gene is part of a p53 target gene cluster that includes GLIPR1L1 and GLIPR1L2, two highly homologous proteins whose expression patterns vary.
Specifications
| GLIPR1 | |
| Polyclonal | |
| Unconjugated | |
| GLIPR1 | |
| GLIPR1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-140 of human GLIPR1 (NP_0068422). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1.35 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| 2410114O14Rik; CRISP7; GLI pathogenesis related 1; GLI pathogenesis-related 1; GLI pathogenesis-related 1 (glioma); glioma pathogenesis-related protein 1; GLIPR; gliPR 1; Glipr1; mRTVP-1; protein RTVP-1; related to testes-specific, vespid, and pathogenesis proteins; related to testis-specific, vespid, and pathogenesis proteins 1; RTVP1; RTVP-1; testes-specific vespid and pathogenesis protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 299783 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction