missing translation for 'onlineSavingsMsg'
Learn More

Glipizide, MedChemExpress

Product Code. 16428196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428196 100 mg 100mg
16412076 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428196 Supplier Medchem Express Supplier No. HYB0254
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Glipizide (CP 2872; K 4024) a potent, orally active and sulfonylurea class anti-diabetic agent and can be used for type 2 diabetes mellitus research but not type 1. Glipizide acts by partially blocking ATP-sensitive potassium (KATP) channels among β cells of pancreatic islets of Langerhans.

Glipizide (CP 2872

Specifications

Chemical Name or Material Glipizide
CAS 29094-61-9
Color White
Purity Grade Notes Research
Molecular Formula C21H27N5O4S
Quantity 100 mg
Synonym CP 28720 K 4024
Solubility Information DMSO : 50 mg/mL (112.22 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.50 mM; Need ultrasonic)
SMILES O=C(C1=NC=C(C)N=C1)NCCC2=CC=C(S(=O)(NC(NC3CCCCC3)=O)=O)C=C2
Molecular Weight (g/mol) 445.54
Formula Weight 445.54
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.