missing translation for 'onlineSavingsMsg'
Learn More

Glipizide-d11, TRC

Product Code. 30390295
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390295 1 mg 1mg
30397287 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30390295 Supplier Toronto Research Chemicals Supplier No. TRCG4102271MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glipizide-d11
CAS 1189426-07-0
Purity Grade Notes HPLC
Molecular Formula C21 D11 H16 N5 O4 S
Quantity 1 mg
Synonym 2-Pyrazinecarboxamide, N-[2-[4-[[[(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl- (ACI), N-[2-[4-[[[(Cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl-2-pyrazinecarboxamide (ACI), Glipizide D11 (cyclohexyl D11)
InChI Formula InChI=1S/C21H27N5O4S/c1-15-13-24-19(14-23-15)20(27)22-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,22,27)(H2,25,26,28)/i2D2,3D2,4D2,5D2,6D2,17D
SMILES [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(NC(=O)NS(=O)(=O)c2ccc(CCNC(=O)c3cnc(C)cn3)cc2)C([2H])([2H])C1([2H])[2H]
IUPAC Name 5-methyl-N-[2-[4-[(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]pyrazine-2-carboxamide
Molecular Weight (g/mol) 456.603
Formula Weight 456.247
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.