missing translation for 'onlineSavingsMsg'
Learn More

Glipizide, 98+%

Product Code. 10768692 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10768692 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10768692 Supplier Thermo Scientific Acros Supplier No. 329110010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 29094-61-9 | C21H27N5O4S | 445.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29094-61-9
Color White
Ignition Residue 0.4% max.
Loss on Drying 1% max. (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C21H27N5O4S
MDL Number MFCD00072159
Quantity 1 g
Merck Index 15, 4477
Synonym glipizide, glucotrol, glydiazinamide, melizide, glibenese, glucozide, glupizide, sucrazide, dipazide, glupitel
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso and methanol
InChI Key ZJJXGWJIGJFDTL-UHFFFAOYSA-N
SMILES CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC Name N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
Molecular Weight (g/mol) 445.5
PubChem CID 3478
ChEBI CHEBI:5384
Formula Weight 445.5
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Glipizide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.