missing translation for 'onlineSavingsMsg'
Learn More

Glimepiride, Thermo Scientific™

A sulfonylurea hypoglecemic agent and potent potassium channel blocker

£64.59 - £256.00

Chemical Identifiers

CAS 93479-97-1
Molecular Formula C24H34N4O5S
Molecular Weight (g/mol) 490.62
MDL Number MFCD00878417
InChI Key WIGIZIANZCJQQY-UHFFFAOYSA-N
Synonym glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin
PubChem CID 3476
IUPAC Name 3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide
SMILES CCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15484489
SDS View Documents
Thermo Scientific Alfa Aesar
J62032.MC
100mg
ONLINE EXCLUSIVE PRICE
£66.00
£64.59 / 100mg
Save £1.41
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15474489
SDS View Documents
Thermo Scientific Alfa Aesar
J62032.03
1g
ONLINE EXCLUSIVE PRICE
£262.00
£256.00 / 1g
Save £6.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Glimepiride induces the PI3 kinase (PI3K) and Akt pathway, along with insulin receptor substrate-1/2 and endothelial nitric oxide synthase. Glimepiride also increases osteoblast proliferation and differentiation, which is thought to be related to its ability to activate the PI3K and Akt pathway. Furthermore, Glimepiride enhances intrinsic peroxisome proliferator-activated receptor γ activity. Glimepiride also increases protein expression of glucose transports 1 and 4, and is a potent KIR channel blocker. Potent Kir6 (KATP) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac Kir6 channels with an IC50 of 6.8 nM.

Solubility
Soluble in DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

93479-97-1
490.62
WIGIZIANZCJQQY-UHFFFAOYSA-N
3476
CCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O
C24H34N4O5S
MFCD00878417
glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin
3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide

Specifications

93479-97-1
MFCD00878417
glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin
WIGIZIANZCJQQY-UHFFFAOYSA-N
3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide
3476
Glimepiride
C24H34N4O5S
14,4440
Soluble in DMSO
CCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O
490.62
490.62
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : UX9363950

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.