missing translation for 'onlineSavingsMsg'
Learn More

Glimepiride, MedChemExpress

Product Code. p-200098438
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16462049

Brand: Medchem Express HYB0104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glimepiride (Glimperide) is a medium-to-long acting sulfonylurea anti-diabetic compound with an ED50 of 182 μg/kg.

Glimepiride (Glimperide) is a medium-to-long acting sulfonylurea anti-diabetic compound with an ED50 of 182 μg/kg.

Specifications

Chemical Name or Material Glimepiride
CAS 93479-97-1
Color White
Purity Grade Notes Research
Molecular Formula C24H34N4O5S
Quantity 1 g
Synonym Glimperide HOE-490
Solubility Information DMSO : 33.33 mg/mL (67.93 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.20 mM; Need ultrasonic)
SMILES O=C(N1C(C(CC)=C(C)C1)=O)NCCC2=CC=C(S(=O)(NC(N[C@H]3CC[C@H](C)CC3)=O)=O)C=C2
Molecular Weight (g/mol) 490.62
Formula Weight 490.62
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.