missing translation for 'onlineSavingsMsg'
Learn More

Gliclazide, TRC

Product Code. 30012765
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012765 1 g 1g
30011770 5 g 5g
30005228 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012765 Supplier Toronto Research Chemicals Supplier No. TRCG4098751G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21187-98-4
Molecular Formula C15 H21 N3 O3 S
Molecular Weight (g/mol) 323.41
Synonym Gliclazide, N-[[(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)amino]carbonyl]-4-methylbenzenesulfonamide, 1-(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)-3-(p-tolylsulfonyl)urea, Cyclopenta[c]pyrrole benzenesulfonamide deriv., 1-(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)-3-(p-tolylsulfonyl)urea, Diabezidum, Diabrezide, Diamicron, Diaprel, Glimicron, Glinormax, Glyzide, N-(4-Methylbenzenesulfonyl)-N'-[3-azabicyclo(3,3,0)oct-3-yl]urea, S 1702, S 852, SE 1702
IUPAC Name 1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-(4-methylphenyl)sulfonylurea
SMILES Cc1ccc(cc1)S(=O)(=O)NC(=O)NN2CC3CCCC3C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H21N3O3S/c1-11-5-7-14(8-6-11)22(20,21)17-15(19)16-18-9-12-3-2-4-13(12)10-18/h5-8,12-13H,2-4,9-10H2,1H3,(H2,16,17,19)
Formula Weight 323.1304
Chemical Name or Material Gliclazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.