missing translation for 'onlineSavingsMsg'
Learn More

Gliclazide, MedChemExpress

Product Code. 16462269
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462269 1 g 1g
16411058 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462269 Supplier Medchem Express Supplier No. HYB0753
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Gliclazide (S1702) is a whole-cell beta-cell ATP-sensitive potassium currents blocker with an IC50 of 184 nM. Gliclazide is used as an antidiabetic.

Gliclazide (S1702) is a whole-cell beta-cell ATP-sensitive potassium currents blocker with an IC50 of 184 nM. Gliclazide is used as an antidiabetic.

Specifications

Chemical Name or Material Gliclazide
CAS 21187-98-4
Color White
Purity Grade Notes Research
Molecular Formula C15H21N3O3S
Quantity 1 g
Synonym S1702 SE1702
Solubility Information DMSO : ≥ 100 mg/mL (309.21 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC=C(C)C=C1)(NC(NN2CC3C(CCC3)C2)=O)=O
Molecular Weight (g/mol) 323.41
Formula Weight 323.41
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.