missing translation for 'onlineSavingsMsg'
Learn More

Glibenclamide, MedChemExpress

Product Code. p-200096750
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16431489

Brand: Medchem Express HY15206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glibenclamide (Glyburide) is an orally active ATP-sensitive K+ channel (KATP) inhibitor and can be used for the research of diabetes and obesity.

Glibenclamide inhibits P-glycoprotein. Glibenclamide directly binds and blocks the SUR1 subunits of KATP and inhibits the cystic fibrosis transmembrane conductance regulator protein (CFTR). Glibenclamide interferes with mitochondrial bioenergetics by inducing changes on membrane ion permeability. Glibenclamide can induce autophagy.

Specifications

Chemical Name or Material Glibenclamide
CAS 10238-21-8
Color White
Purity Grade Notes Research
Molecular Formula C23H28ClN3O5S
Quantity 1 g
Synonym Glyburide
Solubility Information DMSO : 250 mg/mL (506.07 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.20 mM; Need ultrasonic and warming)
SMILES ClC1=CC(C(NCCC2=CC=C(S(NC(NC3CCCCC3)=O)(=O)=O)C=C2)=O)=C(OC)C=C1
Molecular Weight (g/mol) 494.0
Formula Weight 494.0
Percent Purity 98.89%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.