missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glibenclamide, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Glyburide |
| CAS | 10238-21-8 |
| Analyte or Component Names | Glyburide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H28 Cl N3 O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzamide, 5-chloro-N-[2-[4-[[[(cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxy-, Urea, 1-[[p-[2-(5-chloro-o-anisamido)ethyl]phenyl]sulfonyl]-3-cyclohexyl-, 1-[4-[2-(5-Chloro-2-methoxybenzamido)ethyl]phenylsulfonyl]-3-cyclohexylurea, 1-[[-p-[2-(5-Chloro-o-anisamido)ethyl]phenyl]sulfonyl]-3-cyclohexylurea, 1-[p-2-(5-Chloro-o-anisamido)ethylphenylsulfonyl]-3-cyclohexylurea, Abbenclamide, Adiab, Antibet, Apo-Glibenclamide, Azuglucon, Bastiverit, Benclamin, Betanase, Betanaz, Betanese 5, Calabren, Cytagon, Daonil, Daonil N, Debtan, Dia-basan, Diaben, Diabeta, Diabiphage, Dibelet, Duraglucon, Euglucan, Euglucon, Euglucon 5, Euglykon, GBN 5, Gilemal, Gl, Glamide, Gliban, Gliben, Gliben-Puren N, Glibenclamide, Glibenil, Glibens, Glibesyn, Glibet, Glibetic, Glibil, Gliboral, Glicem, Glidiabet, Glimel, Glimide, Glimidstada, Glisulin, Glitisol, Glubate, Gluben, Glucal, Gluco-Tabinen, Glucobene, Glucohexal, Glucolon, Glucomid, Glucoremed, Glucoven, Glukovital, Glyben, Glybenzcyclamide, Glyburide, Glycolande, Glycolande N, Glycomin, Glynase, HB 419, HD 419, Hemi-Daonil, Humedia, Libanil, Maninil, Med-Glionil, Melix, Micronase, Miglucan, N-4-[2-(5-Chloro-2-methoxybenzamido)ethyl]phenylsulfonyl-N'-cyclohexylurea, N-[4-(b-(2-Methoxy-5-chlorobenzamido)ethyl)benzosulfonyl]-N'-cyclohexylurea, N-[4-[b-(2-Methoxy-5-chlorobenzamido)ethyl]benzenesulfonyl]-N'-cyclohexylurea, Norboral, Orabetic, Pira, Prodiabet, Renabetic, Semi-Daonil, Semi-Euglucon, Semi-Euglucon N, Sugril, Suraben, Tiabet, U 26452, UR 606, Wuglucon, Yuglucon, N-[4-(b-(2-Methoxy-5-chlorobenzamido)ethyl)benzosulfonyl]-N'-cyclohexylurea, N-[4-[b-(2-Methoxy-5-chlorobenzamido)ethyl]benzenesulfonyl]-N'-cyclohexylurea |
| InChI Formula | InChI=1S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29) |
| Solution Type | Neat |
| SMILES | COc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction