missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLI3 Recombinant Rabbit Monoclonal Antibody (23GB1280), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GLI3 |
|---|---|
| Clone | 23GB1280 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088120
|
Invitrogen™
MA553126 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088350
|
Invitrogen™
MA553127 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLI3 Recombinant Monoclonal antibody specifically detects GLI3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GLI3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P10071, Q61602 | |
| 140588, 14634, 2737 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1280 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ACLS; add; AI854843; anterior digit pattern deformity; AU023367; Bph; brachyphalangy; extra toes; GCPS; GLI family zinc finger 3; GLI family zinc finger 3 L homeolog; gli3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full length protein; GLI3 full-length protein; gli3.L; GLI3-190; GLI3-83; GLI3FL; GLI-Kruppel family member 3; GLI-Kruppel family member GLI3; GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome); glioma-associated oncogene family zinc finger 3; im:7142603; neural specific DNA binding protein; neural-specific DNA-binding protein xGLI3; oncogene GLI3; PAPA; PAP-A; PAPA1; PAPB; Pdn; PHS; polydactyly Nagoya; PPDIV; Transcriptional activator GLI3; Transcriptional repressor GLI3R; XELAEV_18031191mg; xgli3; xGLI-3; Xt; Zinc finger protein GLI3; zinc finger protein GLI3; transcriptional activator GLI3; zinc finger transcription factor Gli3 | |
| A synthesized peptide derived from human Gli3 (1301-1580AA). | |
| Primary | |
| -20°C | |
| GLI3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title