Learn More
Invitrogen™ GLI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593021
Description
Positive Samples: Mouse lung, Mouse brain, Mouse heart, Mouse ovary, Mouse pancreas, Rat brain; Cellular Location: Cytoplasm, Nucleus Immunogen sequence: METSAPAPAL EKKEAKSGLL EDSSFPDPGK KACPLAVAAA VAAHGVPQQL LPAFHAPLPI DMRHQEGRYH YDPHSVHSVH GPPTLSGSPV ISDISLIRLS PHPAGPGESP FSAHHPYVNP HMEHYLRSVH SSPTLSMISA ARGLSPADVA HEHLKERGLF SLAAPGTNPS DYYHQ.
Gli-2 belongs to the C2H2-type zinc finger protein subclass of the Gli family. Members of this subclass are characterized as transcription factors which bind DNA through zinc finger motifs. These motifs contain conserved H-C links. Gli family zinc finger proteins are mediators of Sonic hedgehog (Shh) signaling and they are implicated as potent oncogenes in the embryonal carcinoma cell. The protein encoded by this gene localizes to the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. The encoded protein is associated with several phenotypes- Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, postaxial polydactyly types A1 and B.
Specifications
| GLI2 | |
| Polyclonal | |
| Unconjugated | |
| GLI2 | |
| AW546128; CJS; GLI family zinc finger 2; GLI family zinc finger protein 2; GLI2; GLI-Kruppel family member GLI2; glioma-associated oncogene family zinc finger 2; HPE9; oncogene GLI2; PHS2; Tax helper protein; tax helper protein 1; tax helper protein 2; tax-responsive element-2 holding protein; tax-responsive element-25-bp sequence binding protein; Thp; THP1; THP2; zinc finger protein GLI2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14633, 304729 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q0VGT2 | |
| GLI2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-175 of mus musculus GLI2 (NP_0010745941). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.