missing translation for 'onlineSavingsMsg'
Learn More

GLI-3 Rabbit anti-Human, Mouse, Rat, Clone: 8B1E2, Novus Biologicals™

Product Code. 18353382 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353382 100 μg 100µL
18312705 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18353382 Supplier Novus Biologicals Supplier No. NBP316166100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GLI-3 Monoclonal antibody specifically detects GLI-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLI-3
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 8B1E2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1301-1580 of human GLI-3 (P10071).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2737
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.