missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-3 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29627PE
This item is not returnable.
View return policy
Description
GLI-3 Polyclonal specifically detects GLI-3 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| GLI-3 | |
| Polyclonal | |
| Flow Cytometry, Flow (Intracellular) | |
| ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| GLI3 | |
| A synthetic peptide made to an internal portion of human Gli3 protein (between residues 50-120) [UniProt P10071] | |
| 0.1 mL | |
| Cancer, Stem Cell Signaling Pathway | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction