missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GLI-3 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 2737 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-29627. For further blocking peptide related protocol, click here . |
| Gene Alias | ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3 |
| Gene Symbol | GLI3 |
| Quantity | 0.1 mg |
| Research Category | Cancer, Stem Cell Signaling Pathway |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?