missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-600R
This item is not returnable.
View return policy
Description
GLI-1 Polyclonal specifically detects GLI-1 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular), Knockdown Validated.
Specifications
| GLI-1 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular), Knockdown Validated | |
| P08151 | |
| GLI1 | |
| A synthetic peptide made toward the C-terminus portion of the human GLI-1 protein (between residues 800-850) [Uniprot: P08151] | |
| 0.1 mL | |
| Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| 2735 | |
| Human, Mouse, Canine | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot, KnockDown | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is directed against Gli1 from mouse brain. No reaction occurs with human or mouse Gli2 or Gli3. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction