missing translation for 'onlineSavingsMsg'
Learn More

Glesatinib hydrochloride, MedChemExpress

Product Code. 16497656
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497656 100 mg 100mg
16480298 5 mg 5mg
16471839 1 mg 1mg
16453787 50 mg 50mg
16437838 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497656 Supplier Medchem Express Supplier No. HY19642A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glesatinib hydrochloride (MGCD265 hydrochloride) is an orally active, potent MET/SMO dual inhibitor. Glesatinib hydrochloride, a tyrosine kinase inhibitor, antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC).

Glesatinib hydrochloride (MGCD265 hydrochloride) is an orally active, potent MET/SMO dual inhibitor. Glesatinib hydrochloride, a tyrosine kinase inhibitor, antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC).

Specifications

Chemical Name or Material Glesatinib hydrochloride
CAS 1123838-51-6
Color Pink
Purity Grade Notes Research
Molecular Formula C31H28ClF2N5O3S2
Quantity 100 mg
Synonym MGCD265 hydrochloride
Solubility Information DMSO : 50 mg/mL (76.20 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC(NC1=CC=C(OC2=C3C(C=C(C4=NC=C(CNCCOC)C=C4)S3)=NC=C2)C(F)=C1)=S)CC5=CC=C(F)C=C5.Cl
Molecular Weight (g/mol) 656.17
Formula Weight 656.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.