missing translation for 'onlineSavingsMsg'
Learn More

GLE1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18433003 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433003 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433003 Supplier Novus Biologicals Supplier No. NBP176975AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLE1 Polyclonal antibody specifically detects GLE1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLE1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias GLE1 (yeast homolog)-like, RNA export mediator, GLE1 RNA export mediator homolog (yeast), GLE1LGLE1 RNA export mediator-like (yeast), GLE1-like protein, GLE1-like, RNA export mediator, hGLE1GLE1 RNA export mediator (yeast), LCCS, LCCS1, lethal congenital contracture syndrome 1, nucleoporin GLE1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human Gle1. The immunogen is located within amino acids 540 - 590 of Gle1. Amino Acid Squence: VKDSKVEQQDNFLKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2733
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.