Learn More
Invitrogen™ GLD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589106
Beschreibung
Immunogen sequence: MFPNSILGRP PFTPNHQQHN NFFTLSPTVY SHQQLIDAQF NFQNADLSRA VSLQQLTYGN VSPIQTSASP LFRGRKRLSD EKNLPLDGKR QRFHSPHQEP TVVNQIVPLS GERRYSMPPL FHTHYVPDIV RCVPPFREIA FLEPREITLP EAKDKLSQQI LELFETCQQQ ISDLKKKELC RTQLQREIQL LFPQSRLFLV GSSLNGFGTR SSDGDLCLVV KEEPCFFQVN QKTEARHILT LVHKHFCTRL SGYIERPQLI RAKVPIVKFR DKVSCVEFDL NVNNIVGIRN TFLLRTYAYL; Positive Samples: U-87MG, BxPC-3, Mouse heart, Rat heart; Cellular Location: Cytoplasm, Nucleus.
GLD2 is cytoplasmic poly(A) RNA polymerase that adds successive AMP monomers to the 3'-end of specific RNAs, forming a poly(A) tail. In contrast to the canonical nuclear poly(A) RNA polymerase, it only adds poly(A) to selected cytoplasmic mRNAs. GLD2 does not play a role in replication-dependent histone mRNA degradation.
Spezifikation
| GLD2 | |
| Polyclonal | |
| Unconjugated | |
| Papd4 | |
| 8030446C20Rik; AI649009; cytoplasmic poly(A) polymerase; Gld2; GLD-2; hGLD-2; hypothetical protein LOC553626; MGC138008 protein; mGLD-2; Palp4; PAP associated domain containing 4; PAP-associated domain-containing protein 4; PAPD4; poly(A) RNA polymerase D4, non-canonical; Poly(A) RNA polymerase GLD2; Tent2; Terminal nucleotidyltransferase 2; terminal uridylyltransferase 2; TUT2; TUTase 2; TUTase2; zgc:110560 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100715, 167153, 361878 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5U315, Q6PIY7, Q91YI6 | |
| Papd4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PAPD4 (NP_7761582). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.