Learn More
Thermo Scientific™ GLD Pro Gas Leak Detector
Product Code. 10438494 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Gas Chromatography Systems
Unit Size:
Each
Product Code. 10438494
Brand: Thermo Scientific™ 66002001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Quickly locate and identify gas leaks in your GC instrument with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Detect and identify gas leaks quickly with the Thermo Scientific™ GLD Pro Gas Leak Detector. Specifically designed for use with GC systems, it detects minute leaks of a wide range of laboratory gases. This hand-held, easy-to-use detector features both visual and audible alarms and utlilizes an LED light display to indicate the intensity of the leak. The small probe detector is designed to reach difficult and confined locations.
Additional Product Information
  • Pushbutton on/off switch
  • Pushbutton zero function
  • Automatic shutoff (five minutes)
  • Rechargable battery (up to six hours operation)
  • Durable storage case
  • Probe holder
  • One-year warranty
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) GC
For Use With (Equipment) Gas Chromatography Systems
Description GLD Pro Gas Leak Detector; Auto shutoff; 6-hr rechargable battery
Type Gas Leak Detector