Learn More
Thermo Scientific™ GLD Pro Gas Leak Detector
Product Code. 17344871 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Gas Chromatography Systems
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17344871 Gas Chromatography Systems Each
1 options
Product Code. 17344871 Supplier Thermo Scientific™ Supplier No. 66002004
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Quickly locate and identify gas leaks in your GC instrument with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Detect and identify gas leaks quickly with the Thermo Scientific™ GLD Pro Gas Leak Detector. Specifically designed for use with GC systems, it detects minute leaks of a wide range of laboratory gases. This hand-held, easy-to-use detector features both visual and audible alarms and utilizes an LED light display to indicate the intensity of the leak. The small probe detector is designed to reach difficult and confined locations.
Additional Product Information
  • Pushbutton on/off switch
  • Pushbutton zero function
  • Automatic shutoff (five minutes)
  • Rechargable battery (up to six hours operation)
  • Durable storage case
  • Probe holder
  • One-year warranty
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) GC
For Use With (Equipment) Gas Chromatography Systems
Description GLD Pro Gas Leak Detector
Product Type Gas Leak Detector