missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLB1L3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48716
This item is not returnable.
View return policy
Description
GLB1L3 Polyclonal antibody specifically detects GLB1L3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GLB1L3 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| beta-galactosidase-1-like protein 3, EC 3.2.1, EC 3.2.1.-, EC 3.2.1.23, FLJ90231, galactosidase, beta 1 like 3, galactosidase, beta 1-like 3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LSGHTKGVLAAINLQKLHQDTFNQLHKVQGASIDAGLLSDASQAATSSSL | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 112937 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction