missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLAST Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20785-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.Specifications
| GLAST | |
| Polyclonal | |
| Unconjugated | |
| SLC1A3 | |
| EA6, EAAT1, GLAST, GLAST 1, GLAST1, SLC1A3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20512, 29483, 6507 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P24942, P43003, P56564 | |
| SLC1A3 | |
| GLAST Fusion Protein Ag14177 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction