missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon splinter powder, 80-200 micron, type 2

Product Code. 11324347
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324347 50 g 50g
11314347 10 g 10g
11304347 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324347 Supplier Thermo Scientific Alfa Aesar Supplier No. 038013.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.

Glassy carbon powder is widely used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Packaging Glass bottle
Quantity 50 g
UN Number UN1325
Solubility Information Insoluble in water.
Chemical Name or Material Glassy carbon splinter powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.