missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon rod, 5mm (0.2in) dia, type 2, Thermo Scientific Chemicals

Widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.

£204.00 - £820.00

Chemical Identifiers

CAS 7440-44-0
MDL Number MFCD00133992
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354307
View Documents Promotion Details
Thermo Scientific Alfa Aesar
037998.DJ
50 mm
£204.00
50mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11364307
View Documents Promotion Details
Thermo Scientific Alfa Aesar
037998.DM
100 mm
£432.00
100mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15401477
View Documents Promotion Details
Thermo Scientific Alfa Aesar
037998.DR
200 mm
£820.00
200mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-44-0
MFCD00133992

Specifications

7440-44-0
MFCD00133992
Glassy carbon rod
Tube
Insoluble in almost all solvents.
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-153-3

missing translation for 'rtecsNumber' : FF5250100

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only