missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon rod, 5mm (0.2 in.) dia, type 2, Alfa Aesar™

Widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.

Brand:  Alfa Aesar 037998.DR

Code : NEW

Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg

 View more versions of this product

Product Code. 15401477

  • £704.00 / 200mm

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Glassy carbon rod
200mm
MFCD00133992
≈0.28g/cm
Tube
7440-44-0
Insoluble in almost all solvents.
Videos
SDS Documents
Certificates