missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glassy carbon foam, nominally 500 pores per inch; Length (in),6 in.; Width (in), 6 in.; Thickness (in), 1 in., Alfa Aesar™
Click to view available options
Quantity:
1 Ea.
Packaging:
Shrink-wrapped
Unit Size:
Each
Specifications
Specifications
| Packaging | Shrink-wrapped |
| MDL Number | MFCD00133992 |
| Quantity | 1 Ea. |
| Chemical Name or Material | Glassy carbon foam |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction