Learn More
Hecht Karl™ Glass Volumetric Pipet
Product Code. 10400002
Change view
Click to view available options
Volume (Metric):
25 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
10400002 25 mL Each
1 options
Product Code. 10400002 Supplier Hecht Karl™ Supplier No. 42550068
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Calibrated Yes
Certifications/Compliance DIN EN ISO 648
Material Glass
Graduated Yes
Graduation Color Amber
Quantity 1 Pack
Tolerance 0.045 mL
Volume (Metric) 25 mL
Type Volumetric Pipet