Learn More
Hirschmann™ Class A Volumetric Flasks with Hollow Glass Stopper
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Hollow Glass Stopper are manufactured from durable DURAN™ glass tubing.
£39.24 - £301.00
Specifications
Certifications/Compliance | DIN EN ISO 1042 |
---|---|
Closure Material | Glass |
Material | Borosilicate Glass, Duran Glass |
Graduated | Yes |
Class | A |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
10597251
|
Hirschmann™
2820292 |
1000 mL | 19/32 NS Standard Ground |
£61.05
Each |
Estimated Shipment: 15-04-2024
Log in to see stock. |
||||
10029760
|
Hirschmann™
2820294 |
2000 mL | 29/32 NS Standard Ground |
£79.95
Each |
Estimated Shipment: 15-04-2024
Log in to see stock. |
||||
11766762
|
Hirschmann™
2820281 |
100 mL | 14/23 NS Standard Ground |
£48.05
Pack of 2 |
In Stock
Log in to see stock. |
||||
11730984
|
Hirschmann™
2820280 |
100 mL | 12/21 NS Standard Ground |
£47.45
Pack of 2 |
Estimated Shipment: 15-04-2024
Log in to see stock. |
||||
11710984
|
Hirschmann™
2820261 |
10 mL | 10/19 NS Standard Ground |
£39.24
Pack of 2 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||||
11726762
|
Hirschmann™
2820260 |
10 mL | 7/16 NS Standard Ground |
£39.25
Pack of 2 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10180851
|
Hirschmann™
2820298 |
10 L | 45/40 NS Standard Ground |
£301.00
Each |
Estimated Shipment: 15-04-2024
Log in to see stock. |
||||
11776762
|
Hirschmann™
2820285 |
200 mL | 14/23 NS Standard Ground |
£53.45
Pack of 2 |
In Stock
Log in to see stock. |
||||
11736762
|
Hirschmann™
2820265 |
20 mL | 10/19 NS Standard Ground |
£42.35
Pack of 2 |
In Stock
Log in to see stock. |
||||
11864412
|
Hirschmann™
2820266 |
20 mL | 12/21 NS Standard Ground |
£40.10
Pack of 2 |
Estimated Shipment: 16-04-2024
Log in to see stock. |
||||
11716063
|
Hirschmann™
2820286 |
250 mL | 14/23 NS Standard Ground |
£54.90
Pack of 2 |
Estimated Shipment: 11-04-2024
Log in to see stock. |
||||
11824272
|
Hirschmann™
2820271 |
25 mL | 12/21 NS Standard Ground |
£46.15
Pack of 2 |
Estimated Shipment: 15-04-2024
Log in to see stock. |
||||
11746762
|
Hirschmann™
2820270 |
25 mL | 10/19 NS Standard Ground |
£40.10
Pack of 2 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||||
10777081
|
Hirschmann™
2820297 |
5 L | 34/35 NS Standard Ground |
£173.00
Each |
Estimated Shipment: 17-04-2024
Log in to see stock. |
||||
11726063
|
Hirschmann™
2820290 |
500 mL | 19/26 NS Standard Ground |
£66.00
Pack of 2 |
Estimated Shipment: 11-04-2024
Log in to see stock. |
||||
11756762
|
Hirschmann™
2820275 |
50 mL | 12/21 NS Standard Ground |
£43.20
Pack of 2 |
Estimated Shipment: 12-04-2024
Log in to see stock. |
||||
11790964
|
Hirschmann™
2820253 |
5 mL | 10/19 NS Standard Ground |
£39.25
Pack of 2 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||||
11720984
|
Hirschmann™
2820278 |
50 mL | 14/23 NS Standard Ground |
£44.25
Pack of 2 |
Estimated Shipment: 15-04-2024
Log in to see stock. |
||||
11716762
|
Hirschmann™
2820252 |
5 mL | 7/16 NS Standard Ground |
£39.25
Pack of 2 |
Estimated Shipment: 25-04-2024
Log in to see stock. |
||||
10534062
|
Hirschmann™
2820293 |
1000 mL | 24/29 NS Standard Ground |
£56.05
Each |
Estimated Shipment: 18-04-2024
Log in to see stock. |
||||
Description
- With NS ground, hollow glass stopper
- With blue stain graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
DIN EN ISO 1042 | |
Borosilicate Glass, Duran Glass | |
A |
Glass | |
Yes |