missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Class A Volumetric Flasks with Rim

Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Rim are manufactured from durable DURAN™ glass tubing.

Brand:  Hirschmann™ 2800197

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10430041

  • £145.00 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

  • With blue graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications

Specifications

DIN EN ISO 1042
Borosilicate Glass, Duran Glass
±1.2 mL
Yes
5 L
470 mm
±1.2 mL
A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers