missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Glass Sample Vials

Product Code. 10099699 Shop All Mettler Toledo Products
Change view
Click to view available options
Volume (Metric):
20 mL
Unit Size:
Pack of 46
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10099699 20 mL Pack of 46
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10099699 Supplier Mettler Toledo™ Supplier No. 51108691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable glass sample vials for the Stromboli KF Oven Sample Changer. Mettler Toledo™ Glass Sample Vials have a 20mL capacity and come in a pack of 46.

  • Suitable for the Stromboli KF Oven Sample Changer
  • Capacity of 20mL
  • 46 vials/pack
TRUSTED_SUSTAINABILITY

Specifications

Material Glass
Volume (Metric) 20 mL
No. per Pack 46
Quantity 46 Pk.
For Use With (Equipment) Stromboli KF Oven Sample Changer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.