Learn More
Hirschmann™ Hollow Glass Stoppers
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
Brand:  Hirschmann™ 8900365
Additional Details : Weight : 0.00450kg
 View more versions of this product
Product Code. 10499631
  • £4.35 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 252
  • Available to fit a range of sizes
Specifications
Specifications
Stopper
Glass
NS 12/21
1/Ea.
For tight sealing of standard ground vessels