Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Product Code. 31781822
Change view
Click to view available options
Volume (Metric):
2 μL
6.66 μL
Unit Size:
Pack of 1000
Pack of 5000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
31781822 2 μL Pack of 5000
10720841 6.66 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31781822 Supplier Hirschmann™ Supplier No. 9000202
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Facilitates error-free capillary filling in the laboratory. Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End are manufactured from durable DURAN™ glass.
  • End-to-end, with minicaps
  • Conformity certified
  • Available either heparinized or non-heparinized
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Accuracy 0.5%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material DURAN
Volume (Metric) 2 μL
Length (Metric) 30 to 32 mm
Type Minicap Capillary Pipette
Disposable Single-use