Learn More
Laboratories Sales™ Glass Snap Top Vials
Brand:  Laboratories Sales™ 105000
Product Code. 11752058
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Manufactured from type 1 neutral glass
  • Particularly suitable for the storage of powders and dry samples
  • Snap ring neck finish fits a flip off style, flexible low density polythene closure supplied separately
  • Single cap fits all vial sizes