missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Flat Tip pH Electrode for Skin and Surfaces with DIN Connector

Product Code. 11632150
Change view
Click to view available options
Connector Types:
7-pin DIN
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
11632150 7-pin DIN Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11632150 Supplier Hanna Instruments™ Supplier No. HI1414D
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Features a built-in temperature sensor for temperature compensated measurements in a single probe design. Hanna Instruments™ Flat Tip pH Electrode for Skin and Surfaces with DIN Connector is a pre-amplified glass body, single reference junction pH electrode made with low temperature glass.

This electrode has a unique open junction design in which there is a hard gel electrolyte layer that is free of silver chloride (AgCl) between the sample and the internal reference cell, resulting in a fast response and stable readings. The open junction design resists clogging and the probe's flat tip makes it ideal for pH measurements on surfaces that cannot be penetrated.

  • Open Junction: Solid gel interface (viscolene) prevents silver from entering the sample and makes it impermeable to clogging
  • Flat Tip: Recommended to optimize surface contact for measurement on skin, leather, agar plates, and paper
  • Built-In Temperature Sensor: Temperature compensated pH measurements without a separate temperature probe
  • Low Temperature Glass: Lower resistance (∼50 megaohms) for use with products/samples stored at low temps (0 to 50°C)
  • Glass Body: Resistant to many harsh chemicals, easily cleaned, and provides fast heat transfer for a steadier reference potential

Specifications

Connector Types 7-pin DIN
Diameter (Metric) 12 mm flat tip
Type pH Electrode
For Use With (Application) For use with HI 99171 Leather and Paper Portable pH Meter
Includes 1m 7-pole cable, internal temperature sensor, amplifier
Length (Metric) 110 mm body, 153.5 mm overall
Material Low-temperature glass body, Viscolene electrolyte
pH Range 0 to 12
Temperature (Metric) Range 0°C to 50°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.