missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Glass Refillable Combination pH Electrode

Product Code. 10486362
Change view
Click to view available options
pH Range:
0 to 13
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
10486362 0 to 13 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10486362 Supplier Hanna Instruments™ Supplier No. HI1131P
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This electrode has a single ceramic junction in the outer reference cell and the spherical pH sensing portion that is made with high temperature glass and is ideal for laboratory samples, liquid samples, and high temperature samples, as well as general purpose use.

  • Features a glass body, refillable, double junction pH electrode with a BNC and pin connector
  • The pin connector is used to enable the CAL Check function of the HI2221, HI2222, and HI2223 benchtop meters
  • This electrode has a single ceramic junction in the outer reference cell and the spherical pH sensing portion is made with high temperature glass
  • This design consideration is ideal for laboratory samples, liquid samples, and high temperature samples, as well as general purpose use

Specifications

Connector Types BNC
Diameter (Metric) 9.5 mm
Type Refillable Combination pH Electrode
For Use With (Application) For general laboratory purposes as well as beer testing
Material Glass
pH Range 0 to 13
Temperature (Metric) Range 20°C to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.