missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Glass Plates (81 x 24 cm), Accessory for SE 600 Ruby and SE 660 Dual Cooled Vertical Electrophoresis Units, Glass Plate
Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
SE 600 Ruby, SE 660 Dual Cooled Vertical Gel Electrophoresis Unit
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | SE 600 Ruby, SE 660 Dual Cooled Vertical Gel Electrophoresis Unit |
| Type | Glass Plate |
| Material | Glass |
| Length (Metric) | 24 cm |
| Width (Metric) | 18 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction