missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Glass Media Bottles

Product Code. 31923917
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
5 L
Unit Size:
Each
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31923917 5 L Each
31923132 50 mL Pack of 10
31921733 100 mL Pack of 10
31921963 250 mL Pack of 10
31922064 500 mL Pack of 10
31923921 1 L Pack of 10
31921501 2 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 31923917 Supplier Globe Scientific Supplier No. 8105000
Upgrade Your Laboratory Essentials with Globe Scientific
Use promo code 27959
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Globe Glass™ media bottles are designed with a wide mouth for ease of filling, dispensing and pipetting.

  • A drip free pour ring and autoclavable polypropylene (PP) screw cap enable sterilizations of media and reagents.
  • Globe Glass™ media bottles are available in sizes 50 mL to 10000 mL.
  • ASTM E438, Type I, Class A, Borosilicate 3.3 Glass
  • GL32 polypropylene (PP) screw cap on size 50 mL
  • GL45 polypropylene (PP) screw caps on sizes 100 mL to 10 L
  • Pouring ring for drip free liquid transfer
  • High resistance to chemicals
  • Dual scale metric graduations
  • Durable white enamel printing
  • Large white marking area

Specifications

Capacity (Metric) 5 L
Color Clear
Material Borosilicate Glass (3.3)
Sterility Non-sterile
Closure Included Yes
Graduated Yes
Mouth Threaded
Closure Size GL45
Autoclavable Autoclavable
Capacity (English) 169.07 oz.
Shape Round
Disposable Reusable
Product Type Media Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.