Learn More
Trajan Scientific and Medical™ Glass Lined Stainless Steel Union
Click to view available options
Diameter (Metric) Inner:
0.5 mm
0.8 mm
Unit Size:
Each
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
Specifications
| Type | Union |
| Material | Stainless Steel |
| Diameter (Metric) Inner | 0.5 mm |