missing translation for 'onlineSavingsMsg'
Learn More

Manutan Glass handle with printed tape, LxW 66m x 50mm

Product Code. 12816019
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12816019 Supplier Manutan Supplier No. A329794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Strong low noise polypropylene tape on a 75mm core. Supplied in cartons of 36 rolls, 1 message.
  • Roll width : 50mm
  • Roll length : 66m
  • Printed message : GLASS HANDLE WITH CARE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.