Learn More
Hirschmann™ Class AS Graduated Pipettes, Blue Graduation
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes, Blue Graduation come with a cotton plug end as standard.
Brand:  Hirschmann™ 1101175
Code : 50
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 11730834
  • £137.00 / Pack of 6
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN EN ISO 835
  • Main point ring graduation in blue
  • Graduated to tip, zero on top, type 3
  • Conformity certified, with dated batch identification
Specifications
Specifications
Yes
AS
Blue
Spare Adapter
DIN EN ISO 835
Silicone
50 mL
Bibbette Pipet Filler
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products