Learn More
Poulten Graf™ Glass BOD Bottle
Product Code. 12397208
Change view
Click to view available options
Volume (Metric):
250 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
12397208 250 mL Each
1 options
Product Code. 12397208 Supplier Poulten Graf™ Supplier No. GS748/A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Prevents air entrapment for your biological oxygen demand needs. Poulten Graf™ Glass BOD Bottle is made of soda lime Volac™ glass with an interchangeable stopper.
  • Glass peg stopper adjusted to leave a "well" for water seal at bottle opening
  • Pointed cone on stopper to prevent air entrapment
  • Manufactured from clear glass
Specifications
Volume (Metric) 250 mL
Material Glass