missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Glass Beads for Mills

For use with Retsch MM 200 Mixer Mills.

Brand:  RETSCH 22.222.0002

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11736175

  • £62.80 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

  • Ideal for disruption of biological cells
Specifications

Specifications

Beads
0.25 to 0.50 mm
500 g
Glass
Mixer Mill MM 200
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers