Learn More
RETSCH Glass Beads for Mills
For use with Retsch MM 200 Mixer Mills.
Brand:  RETSCH 22.222.0003
Additional Details : Weight : 0.45720kg
 View more versions of this product
Product Code. 10262641
  • £80.65 / 500g
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Ideal for disruption of biological cells
Specifications
Specifications
Beads
0.50 to 0.75 mm
500 g
Glass
Mixer Mill MM 200