missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Glass Beads, 2 mm
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Description
- Made of silica glass
- Sample type: Animal Tissues, Yeast, Fungi
- Specific Gravity: 2.55
Specifications
Specifications
| Diameter (English) | 0.07 to 0.08 in. |
| Diameter (Metric) | 1.7 to 2.1 mm |
| For Use With (Equipment) | FastPrep-24 5G; FastPrep-24 Homoginizers |
| For Use With (Application) | Sample preparation, sample lysis, nucleic acid isolation and purification, protein isolation, molecular biology, gene expression, microbiological analysis, gene function, gene regulation |
| Product Type | Silica Glass Beads |
| Quantity | 1 kg |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction