missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Glasgow's MEM
GREENER_CHOICE

Product Code. 11580576
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11580576 10 x 500 mL Pack of 10
11570576 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11580576 Supplier Gibco™ Supplier No. 21710082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Glasgow's MEM

Glasgow's Minimum Essential Medium (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's medium. It was utilized to study the genetic factors which affected cell competence. Glasgow's MEM was developed for use with kidney cells lines, such as BHK-21.

Glasgow's MEM is unique from other media as it contains twice the concentration of amino acids and vitamins than the original Basal Medium Eagle, and is used without serum. Glasgow's MEM was originally formulated with 10% tryptose phosphate broth.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

Dual-Site cGMP Manufacturing
Gibco™ Glasgow's MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Glasgow's MEM product made in our Grand Island facility (11710-035). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Glasgow's MEM contains no proteins, lipids, or growth factors. Therefore, Glasgow's MEM requires supplementation, commonly with 10% tryptose phosphate broth. Glasgow's MEM uses a sodium bicarbonate buffer system (2.75 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Glasgow's MEM medium
With Additives Glutamine, Phenol Red
Without Additives No HEPES
For Use With (Application) Mammalian cell culture
Quantity 10 x 500 mL
Certifications/Compliance ISO 13485 standards, FDA
Regulatory Status For in-vitro diagnostic use
Content And Storage 2 to 8°C
Description Glasgow's MEM Medium, 500mL, Liquid Form, With L-glutamine, Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.