missing translation for 'onlineSavingsMsg'
Learn More

Glasdegib, MedChemExpress

Product Code. p-200100684
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16467096

Brand: Medchem Express HY16391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glasdegib (PF-04449913) is a potent and orally bioavailable smoothened inhibitor. Glasdegib (PF-04449913) binds to human SMO (amino acids 181-787) with an IC50 of 4 nM.

Glasdegib (PF-04449913) is a potent and orally bioavailable smoothened inhibitor. Glasdegib (PF-04449913) binds to human SMO (amino acids 181-787) with an IC50 of 4 nM.

Specifications

Chemical Name or Material Glasdegib
CAS 1095173-27-5
Color White
Purity Grade Notes Research
Molecular Formula C21H22N6O
Quantity 100 mg
Synonym PF-04449913
Solubility Information DMSO : ≥ 83.33 mg/mL (222.55 mM)
SMILES O=C(NC1=CC=C(C#N)C=C1)N[C@H]2C[C@H](C3=NC4=CC=CC=C4N3)N(C)CC2
Molecular Weight (g/mol) 374.44
Formula Weight 374.44
Percent Purity 98.36%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.