missing translation for 'onlineSavingsMsg'
Learn More

Glasdegib, MedChemExpress

Product Code. 16467096
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467096 100 mg 100mg
16437048 10 mg 10mg
16413097 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16467096 Supplier Medchem Express Supplier No. HY16391
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Glasdegib (PF-04449913) is a potent and orally bioavailable smoothened inhibitor. Glasdegib (PF-04449913) binds to human SMO (amino acids 181-787) with an IC50 of 4 nM.

Glasdegib (PF-04449913) is a potent and orally bioavailable smoothened inhibitor. Glasdegib (PF-04449913) binds to human SMO (amino acids 181-787) with an IC50 of 4 nM.

Specifications

Chemical Name or Material Glasdegib
CAS 1095173-27-5
Color White
Purity Grade Notes Research
Molecular Formula C21H22N6O
Quantity 100 mg
Synonym PF-04449913
Solubility Information DMSO : ≥ 83.33 mg/mL (222.55 mM)
SMILES O=C(NC1=CC=C(C#N)C=C1)N[C@H]2C[C@H](C3=NC4=CC=CC=C4N3)N(C)CC2
Molecular Weight (g/mol) 374.44
Formula Weight 374.44
Percent Purity 98.36%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.