Learn More
Ansell™ Gladiator™ 16-650 Series Heavy Weight Gloves
Product Code. p-8005273 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Product Code. 12670772
Brand: Ansell™ 16650080
Only null left
Add to basket
Add to basket
Designed for light-to-medium stress glass handling. Crinkle finish provides excellent wet and dry grip on slippery surfaces but also protects the hand from sharp edges.
  • Crinkle finish for grip on slipper surfaces and protection from sharp edges
  • Natural rubber coating
  • Cut-and-sewn construction
  • Interlock cotton liner
  • Fully-coated finish
  • Gauntlet cuff
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Natural Rubber Latex
Color Green
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material Rubber Latex
Product Type Heavyweight Gloves
Size 8
Finger Style Crinkled finish
For Use With (Application) Glass repair and replacement Construction Utilities All food processing applications that have food contact Seafood processing
Glove Cuff Color Green
Glove Cuff Style Gauntlet
Liner Material Interlock Cotton
Lining Lined
Quantity 12 Pairs
Resistant To Good cushioning, good cut resistance
Series 16-650
Length (English) 12.2
Length (Metric) 310